Learn More
C9orf40 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559900
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VASRQHNEEF WQYNTFQYWR NPLPPIDLAD IEDLSEDTLT EATLQGRNEG AEVDME Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 70%.
The function of C9orf40 remains unknown.Spezifikation
C9orf40 | |
Polyclonal | |
Unconjugated | |
C9orf40 | |
C9orf40; chromosome 9 open reading frame 40; FLJ10110; uncharacterized protein C9orf40 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55071 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXQ3 | |
C9orf40 | |
Recombinant protein corresponding to Human C9orf40 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only