Learn More
Thermo Scientific Chemicals Gen-assoziiertes Calcitonin-Peptid, Ratte
für zell- und molekularbiologische Anwendungen.
Marke: Thermo Scientific Chemicals J66827.|D
Weitere Details : CAS : 96827-03-1 Netto-Gewicht : 0.00001kg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Rat | |
-30°C to -10°C | |
3806.24 | |
0.5 mg | |
CGRP, rat |
HPLC | |
Fest | |
Calcitonin Gene Related Peptide, rat | |
RUO - research use only |
Nur für Forschungszwecke