missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Polyclonal Antibody, Invitrogen™
Beschreibung
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Calnexin, also referred to as IP90, p88 and p90, is an ∽90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
Spezifikation
Spezifikation
| Antigen | Calnexin |
| Anwendungen | Immunofluorescence, Immunohistochemistry, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.85 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA and 0.05% sodium azide; pH 7.4 |
| Gen | CANX |
| Gen-Zugriffsnummer | P27824, P35564, P35565 |
| Gen-Alias | Calnexin |
| Gensymbole | Canx |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?