missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpain-Inhibitor II, ≥ 95 %

Catalog Number p-7082312
Click to view available options
Menge:
5 mg
25 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
136632-32-1
C19H35N3O4S, C19H35N3O4S
N-Ac-Leu-Leu-Methioninal; ALLM
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
136632-32-1
C19H35N3O4S, C19H35N3O4S
N-Ac-Leu-Leu-Methioninal; ALLM

Ein Inhibitor von Calpains I und II

ALLM (Calpain Inhibitor) is a cell-permeable, peptide aldehyde inhibitor of calpain I and calpain II. ALLM acts as a very potent inhibitor of cathepsin L and the strongest inhibitor of cathepsin B amongst the peptide aldehydes. This compound inhibits the processing of malaria aspartic hemoglobinases plasmepsins I and II in vitro. ALLM also inhibits other neutral cysteine proteases, and activation-induced programmed cell death. It restores defective immune responses in HIV+ donors. ALLM blocks nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. ALLM also acts as a weak inhibitor of proteasome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Menge 5 mg
Beschreibung White
Inhalt und Lagerung -30°C to -10°C
Zur Verwendung mit (Anwendung) Ein Inhibitor von Calpains I und II
Formelmasse 401.60g/mol
Löslichkeit Soluble in ethanol at 20mg/ml
Produkttyp Signal Transduction Reagent
Form Pulver
InChI-Schlüssel RJWLAIMXRBDUMH-UHFFFAOYNA-N
SMILES CSCCC(NC(=O)C(CC(C)C)NC(=O)C(CC(C)C)NC(C)=O)C=O
IUPAC-Name 2-acetamido-4-methyl-N-(3-methyl-1-{[4-(methylsulfanyl)-1-oxobutan-2-yl]carbamoyl}butyl)pentanamide
Molekulargewicht (g/mol) 401.57
Assay ≥95%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'