missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CAMK1D (CaMKI Delta), His Tag Recombinant Protein
Human CAMK1D (CaMKI Delta), His Tag Recombinant Protein
Marke: Invitrogen PV3663
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
CAMK1D (CaMK1 delta) is a serine⁄threonine kinase that is a member of the calcium⁄calmodulin-dependent protein kinase family. CAMK1D is expressed in polymorphonuclear leukocytes and may be part of the chemokine signal transduction pathway that regulates granulocyte function. It may also be involved in modulation of neuronal apoptosis.Spezifikation
NP_705718 | |
57118 | |
10 μg | |
CAMK1 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
calcium⁄calmodulin-dependent protein kinase ID | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
47 kDa | |
CAMK1D (CaMKI Delta) | |
Human | |
His Tag | |
Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
CAMK1D | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |