missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Marke: Thermo Scientific™ PV4205
Weitere Details : Netto-Gewicht : 0.05250kg
Beschreibung
As a dominant kinase in the central nervous system CAMK2B (CaMKII beta) may function in long term potentiation and neurotransmitter release.Spezifikation
NP_742078 | |
114294, 816 | |
10 μg | |
CAMK2 Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CAMK Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™ | |
calcium⁄calmodulin-dependent protein kinase (CaM kinase) II beta | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
60.5 kDa | |
CAMK2B (CaMKII Beta), CAMK2 | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
CAMK2B | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |