Learn More
Camostat mesylate, Tocris Bioscience™
Orally active protease inhibitor
Marke:  Tocris (Bio-Techne) 319350
Weitere Details : Netto-Gewicht : 0.01000kg
Menge. 50mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412404
  • 211.20 € / 50 Milligramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Orally active protease inhibitor. Known to inhibit trypsin and various inflammatory proteases including plasmin, kallikrein and thrombin. Suppresses pancreatitis-induced pain in rats following oral administration.
Spezifikation
Chemische Identifikatoren
59721-29-8
494.519
camostat mesylate, camostat mesilate, camostat mesylate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
[4-[2-[2-(dimethylamino)-2-oxoethoxy]-2-oxoethyl]phenyl] 4-(diaminomethylideneamino)benzoate;methanesulfonic acid
C21H26N4O8S
FSEKIHNIDBATFG-UHFFFAOYSA-N
5284360
CN(C)C(=O)COC(=O)CC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)N=C(N)N.CS(=O)(=O)O
Spezifikation
Camostat mesylate
59721-29-8
camostat mesylate, camostat mesilate, camostat mesylate, camostat methanesulfonate, camostat monomethanesulfonate, foipan, unii-451m50a1eq, dsstox_cid_238, dsstox_rid_75452, dsstox_gsid_20238
CN(C)C(=O)COC(=O)CC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)N=C(N)N.CS(=O)(=O)O
494.519
494.52
50mg
C21H26N4O8S
FSEKIHNIDBATFG-UHFFFAOYSA-N
[4-[2-[2-(dimethylamino)-2-oxoethoxy]-2-oxoethyl]phenyl] 4-(diaminomethylideneamino)benzoate;methanesulfonic acid
5284360
>98%
Videos