Learn More
Jaytec Glass™ Capillary Sealing Compound
Product Code. 12396287
Change view
Click to view available options
Material:
Plastic
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12396287 Plastic Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12396287 Supplier Jaytec Glass™ Supplier No. CAPCS10
Verfügbar ab: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Capillary Sealing Compound is used to plug the end of a capillary tube. The kit consists of a non-drying cement in a plastic tray. Push the empty end of a filled capillary tube into the paste to seal it.
  • 24 capillaries can be placed upright in the numbered storage holes on either side of the plate.
Specifications
Material Plastic
Product Type Sealing Compound