Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda Mouse Conjugate
Alle ansehen Thermo Scientific ProdukteDescripción
CaptureSelect Biotin Anti-LC-lambda Mouse Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of lambda light chains of mouse antibodies. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
Applications for CaptureSelect Biotin Anti-LC-lambda Mouse Conjugate include ELISA, western blot, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Especificaciones
Especificaciones
| Inhalt und Lagerung | Store in freezer (-5°C to -30°C). |
| Form | Flüssig |
| Produktlinie | CaptureSelect |
| Menge | 100 μg |
| Typ | Conjugate |
| Marker oder Farbstoff | Biotin & Analogs |
| Ligandentyp | Antikörper |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.