Learn More
CARS Antibody (CL2302) [Janelia Fluor™ 549], Novus Biologicals™
Artikelnummer. 30292387 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30292387 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 30292387 Lieferant Novus Biologicals Lieferanten-Nr. NBP236755JF549
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen CARS
Anwendungen Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon CL2302
Konjugat Janelia Fluor 549
Zusammensetzung 50mM Sodium Borate
Gen-Alias CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246
Wirtsspezies Mouse
Immunogen This antibody was developed using a recombinant protein derived from A8MVQ3, with the exact immunogen sequence remaining proprietary.
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 833
Zielspezies Human
Inhalt und Lagerung Store at 4⪚C in the dark.
Form Purified
Isotype IgG1