Learn More
CASC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA558872
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ESEAIKCERE MKVLSETVSA AQLLLVENSS EKPDFFEDNV VDLCQFTTLG GVYHLDILEL PPQCKPVKGW MIVEILKEGL QKY Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 61%.
CASC1 Polyclonal antibody specifically detects CASC1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
CASC1 | |
Polyclonal | |
Unconjugated | |
CASC1 | |
cancer susceptibility candidate 1; cancer susceptibility candidate gene 1 protein; CASC1; LAS1; lung adenoma susceptibility 1; lung adenoma susceptibility 1-like protein; PPP1R54; Protein CASC1; protein phosphatase 1 regulatory subunit 54; protein phosphatase 1, regulatory subunit 54 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55259 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6TDU7 | |
CASC1 | |
Recombinant protein corresponding to Human CASC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only