missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casc5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA549174
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Casc5 Polyclonal antibody specifically detects Casc5 in Mouse samples. It is validated for Western Blot.Spezifikation
Casc5 | |
Polyclonal | |
Unconjugated | |
KNL1 | |
2310043D08Rik; 5730505K17Rik; AF15Q14; ALL1-fused gene from chromosome 15q14 protein; Blinkin; blinkin, bub-linking kinetochore protein; Bub-linking kinetochore protein; cancer susceptibility candidate 5; cancer susceptibility candidate gene 5 protein; Cancer/testis antigen 29; CASC5; CT29; D40; hKNL-1; hSpc105; KIAA1570; kinetochore null 1 homolog; kinetochore scaffold 1; Kinetochore-null protein 1; KNL1; MCPH4; microcephaly, primary autosomal recessive 4; PPP1R55; Protein CASC5; Protein D40/AF15q14; protein phosphatase 1, regulatory subunit 55; Rad51; RAD51 homolog; Spc7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
76464 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q66JQ7 | |
KNL1 | |
KLH conjugated synthetic peptide between 1557-1583 amino acids from the C-terminal region of mouse Casc5. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |