missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin S Antibody (04), Novus Biologicals™

Artikelnummer. 18690184 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
Unit Size:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18690184 100 μg 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18690184 Supplier Novus Biologicals Supplier No. NBP323489

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin S
Anwendungen ELISA
Klassifikation Monoclonal
Klon 4
Konjugat Unconjugated
Verdünnung ELISA 1:1000-1:2000
Zusammensetzung 0.2 um filtered solution in PBS
Gen-Alias cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Wirtsspezies Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cathepsin S (Accession#: NP_004070.3; Met1-Ile331).
Reinigungsverfahren Protein A purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Immunology
Primär oder sekundär Primary
Gen-ID (Entrez) 1520
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.