Learn More
CATSPER4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA560965
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GQQQRITFSE TGAEEEEEND QLPLVHCVVA RSEKSGLLQE PLAGGPLSNL SENTCDNFCL VLEAIQENLR QYKEIRD Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 71%.
CATSPER4 Polyclonal antibody specifically detects CATSPER4 in Human samples. It is validated for Immunohistochemistry.Spezifikation
CATSPER4 | |
Polyclonal | |
Unconjugated | |
CATSPER4 | |
4933412L05; cation channel of sperm 4; cation channel sperm associated 4; cation channel sperm-associated protein 4; cation channel, sperm associated 4; CatSper4; channel, sperm associated 4; CTSR4; ion channel CatSper4; RGD1565213 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
378807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTX7 | |
CATSPER4 | |
Recombinant protein corresponding to Human CATSPER4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only