Learn More
CCDC155 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA554115
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EENGKLLAER DGVKKRSQEL AMEKDTLKRQ LFECEHLICQ RDTILSERTR DVESLAQTLE EYRVTTQELR LEISRLEEQL SQTYEGPDEL PEGA Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.
CCDC155 Polyclonal antibody specifically detects CCDC155 in Human samples. It is validated for Immunohistochemistry.Spezifikation
CCDC155 | |
Polyclonal | |
Unconjugated | |
CCDC155 | |
CCDC155; coiled-coil domain containing 155; coiled-coil domain-containing protein 155; KASH domain containing 5; KASH domain-containing protein 5; KASH5; protein KASH5; Unknown (protein for MGC:134376) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N6L0 | |
CCDC155 | |
Recombinant protein corresponding to Human CCDC155 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only