missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC62 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25981-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The function of this protein has not been specifically defined.Spezifikation
CCDC62 | |
Polyclonal | |
Unconjugated | |
CCDC62 | |
aaa, CCDC62, Protein aaa, Protein TSP NY, TSP NY | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
208908, 84660 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
E9PVD1, Q6P9F0 | |
CCDC62 | |
CCDC62 Fusion Protein Ag22043 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |