missing translation for 'onlineSavingsMsg'
Learn More

PeproTech CCL5 (RANTES) Polyclonal Antibody, PeproTech®, Invitrogen™

Artikelnummer. p-7245169
Change view
Click to view available options
Menge:
1 mg
100 μg
50 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
50 Mikrogramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17853583 50 μg 50 Mikrogramm
17873483 100 μg 100 Mikrogramm
17853663 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17853583 Lieferant PeproTech Lieferanten-Nr. 500P7850UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat RANTES (CCL5). Anti-Rat RANTES (CCL5)-specific antibody was purified by affinity chromatography employing an immobilized Rat RANTES (CCL5) matrix. Sandwich ELISA: To detect Rat RANTES (CCL5) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat RANTES (CCL5) (500-P78Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat RANTES (CCL5). Western Blot: To detect Rat RANTES by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat RANTES is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CCL5 (RANTES)
Anwendungen ELISA, Western Blot
Klassifikation Polyclonal
Konzentration 0.1-1.0 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen CCL5
Gen-Zugriffsnummer P50231
Gen-Alias beta-chemokine RANTES; CC chemokine; C-C motif chemokine 5; C-C motif chemokine ligand 5; CCL5; chemokine (C-C motif) ligand 5; D17S136E; EoCP; eosinophil chemotactic cytokine; MGC17164; MuRantes; OTTHUMP00000197106; RANTES; RANTES protein; RANTES(3-68); RANTES(4-68); regulated upon activation normal T-cell expressed and secreted; regulated upon activation, normally T-expressed, and presumably secreted; Scya5; SIS δ; SISd; SIS-delta; SISδ; small inducible cytokine A5; small inducible cytokine subfamily A (Cys-Cys), member 5; small-inducible cytokine A5; T cell-specific protein P228; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; Unknown (protein for MGC:127014)
Gensymbole CCL5
Wirtsspezies Rabbit
Immunogen E.coli-derived Recombinant Rat RANTES (CCL5).
Reinigungsverfahren Antigen affinity chromatography
Menge 50 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 81780
Zielspezies Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Lyophilized
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.