Learn More
CCP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16812544 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16812544 20 μL 20µL
16802544 150 μL 150µL
2 options
Product Code. 16812544
Brand: Proteintech 140671AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
NNA1 is a zinc carboxypeptidase that contains nuclear localization signals and an ATP/GTP-binding motif that was initially cloned from regenerating spinal cord neurons of the mouse.
Specifications
Antigen CCP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AGTPBP1
Gene Accession No. Q641K1, Q9UPW5
Gene Alias AGTPBP1, ATP/GTP binding protein 1, CCP1, Cytosolic carboxypeptidase 1, KIAA1035, NNA1
Gene Symbols Agtpbp1
Host Species Rabbit
Immunogen CCP1 Fusion Protein Ag5122
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23287, 67269
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG