missing translation for 'onlineSavingsMsg'
Learn More

CCT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10571-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887033

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CCT3 (Chaperonin Containing TCP1 Subunit 3) is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT). The complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8. Diseases associated with CCT3 include Spinocerebellar Ataxia 10 and Immature Cataract.
Spezifikation

Spezifikation

CCT3
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49368, P80318, Q6P502
CCT gamma, CCT3, TCP 1 gamma
Rabbit
IgG
150 μL
-20°C
Primary
12462, 295230, 7203
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
CCT3
Liquid
CCT3
CCT3 Fusion Protein Ag0843
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate