missing translation for 'onlineSavingsMsg'
Learn More

CD11c Hamster anti-Mouse, APC-R700, Clone: N418, BD Horizon™

Hamster Monoclonal Antibody

Marke:  BD Biosciences 565872

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15804579

  • 510.00 € / 1 Stück
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.

CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CD11c
Monoclonal
0.2mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Hamster
0.1 mg
Cell Biology
Mouse
Purified
Flow Cytometry
N418
APC-R700
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4
Mouse Dendritic Cells
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
IgG2
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.