missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138 Mouse anti-Human, APC-R700, Clone: MI15 , BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566051
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.
CD138 Monoclonal antibody specifically detects CD138 in Human samples. It is validated for Flow cytometry.Spezifikation
CD138 | |
Monoclonal | |
APC-R700 | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
MI15 | |
Vol. Per test: 5μL | |
Syndecan; SDC; Syndecan 1; SDC1; SYND1 | |
Human U266 and XG-1 Myeloma Cell Lines | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. | |
IgG1 κ |
For Research Use Only.