Learn More
CD140a (PDGFRA) Monoclonal Antibody (APA5), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
131.00 € - 342.00 €
Spezifikation
Antigen | CD140a (PDGFRA) |
---|---|
Klon | APA5 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Super Bright 600 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16321454
|
Affymetrix eBioscience
63-1401-82 |
100 μg |
342.00 €
100 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16311454
|
Invitrogen
63-1401-80 |
25 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues. Applications Reported: This APA5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
CD140a (PDGFRA) Monoclonal antibody specifically detects CD140a (PDGFRA) in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD140a (PDGFRA) | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
18595 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
APA5 | |
Monoclonal | |
Liquid | |
RUO | |
P26618 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |