Learn More
CD150 Monoclonal Antibody (mShad150), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
138.00 € - 317.00 €
Spezifikation
Antigen | CD150 |
---|---|
Klon | mShad150 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15903414
|
Affymetrix eBioscience
62-1502-80 |
25 μg |
138.00 €
25 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15913414
|
Affymetrix eBioscience
62-1502-82 |
100 μg |
317.00 €
100 Mikrogramm |
Verfügbar ab: 21-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The mShad150 monoclonal antibody reacts with mouse CD150, an ∽70 kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells. Applications Reported: This mShad150 antibody has been reported for use in flow cytometric analysis. Applications Tested: This mShad150 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent.
CD150 Monoclonal antibody specifically detects CD150 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD150 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q9QUM4 | |
Slamf1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Slamf1 |
mShad150 | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Mouse | |
27218 | |
IgG2b λ | |
Affinity chromatography | |
Antibody |