missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, BUV496, Clone: 3G8, BD Horizon™

Mouse Monoclonal Antibody

Marke:  BD Biosciences 564654

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15803789

  • 270.00 € / 1 Stück

Verfügbar ab
16-10-2020

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD16 Monoclonal antibody specifically detects CD16 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Spezifikation

Spezifikation

CD16
3G8
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c x DBA/2, also known as CD2F1 or CDF1 IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimum conditions, and unconjugated antibody and free BD Horizon BUV496 were removed.
Monoclonal
Flow Cytometry
Brilliant Ultraviolet 496
Aqueous buffered solution containing ≤0.09% sodium azide.
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Human polymorphonuclear leukocytes
25 Tests
Cell Biology
Primary
Baboon, Cynomolgus, Human, Rhesus
Videos
Dokumentation

For Research Use Only.