missing translation for 'onlineSavingsMsg'
Learn More

CD161 Mouse anti-Human, APC, Clone: DX12, BD

Artikelnummer. 15841279
Change view
Click to view available options
Menge:
100 Tests
25 Tests
Packungsgröße:
1 Stück
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15841279 25 Tests 1 Stück
15876448 100 Tests 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15841279 Lieferant BD Biosciences Lieferanten-Nr. 561729

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

The DX12 monoclonal antibody specifically binds to human CD161 which is also known as Natural killer cell surface protein P1A (NKR-P1A). CD161 is encoded by the KLRB1 (Killer cell lectin-like receptor subfamily B member 1) gene. CD161 is a member of the C-type lectin superfamily and is also referred to as C-type lectin domain family 5 member B (CLEC5B). CD161 is a 80 kDa disulfide-linked homodimer, type II membrane glycoprotein. CD161 is expressed mostly on NK cell populations and on subsets of CD4+ and CD8+ αβ T cells, NKT cells and γδ T cells. Reports indicate that CD161 is expressed preferentially on CD45RO+ T cells, however, it can be found on a subset of thymocytes and fetal liver T cells. Its function has not been fully elucidated, but reports indicate that NKR-P1A may serve as a specific receptor for some NK cell targets. The DX12 antibody can reportedly inhibit spontaneous cytotoxicity mediated by certain NK cell clones.

TRUSTED_SUSTAINABILITY

Specifikationer

Antigen CD161
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon DX12
Konjugat APC
Zusammensetzung Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Wirtsspezies Mouse
Immunogen Human NKR-P1A Transfected Mouse Fibroblasts
Reinigungsverfahren Affinity Purified
Menge 25 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Visa mer Visa mindre

For Research Use Only.

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.