Learn More
CD19 Monoclonal Antibody (SJ25C1), Super Bright™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
152.00 € - 352.00 €
Spezifikation
Antigen | CD19 |
---|---|
Klon | SJ25C1 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15904144
|
Affymetrix eBioscience
78-0198-42 |
100 Tests |
352.00 €
100 Stück |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen | Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15994134
|
Invitrogen
78-0198-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The HIB19 monoclonal antibody reacts with human CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes. Applications Reported: This HIB19 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIB19 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 780 nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD19 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P15391 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
SJ25C1 | |
Flow Cytometry | |
Super Bright 780 | |
Mouse | |
Human | |
930 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |