Learn More
CD24 Monoclonal Antibody (M1/69), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
366.00 €
Spezifikation
Antigen | CD24 |
---|---|
Klon | M1/69 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15913614
|
Affymetrix eBioscience
64-0242-82 |
100 μg |
366.00 €
100 Mikrogramm |
Verfügbar ab: 14-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The SN3 monoclonal antibody reacts with the human CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. It has been reported that P-selectin (CD62P) binds to CD24. Applications Reported: This eBioSN3 (SN3 A5-2H10) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioSN3 (SN3 A5-2H10) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD24 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P24807 | |
Cd24a | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd24a |
M1/69 | |
Flow Cytometry | |
Super Bright 645 | |
Rat | |
Mouse | |
12484 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |