Learn More
CD266 (TWEAK Receptor) Monoclonal Antibody (ITEM-1), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
135.00 € - 305.00 €
Spezifikation
Antigen | CD266 (TWEAK Receptor) |
---|---|
Klon | ITEM-1 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16350634
|
Affymetrix eBioscience
12-9019-42 |
100 Tests |
305.00 €
100 Stück |
Verfügbar ab: 15-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16340634
|
Affymetrix eBioscience
12-9019-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The ITEM-1 mAb reacts with human TWEAK Receptor/Fn14 (fibroblast growth factor-inducible 14 kDa protein). Fn14 is distantly related to the TNFR family, containing one cysteine-rich domain in the extracellular region and a TNFR-associated factor binding domain but does not contain a death domain (DD) cytoplasmic region. Fn14 plays a role in TWEAK-induced endothelial cell migration, proliferation, and angiogenesis. TWEAK-induced cell death via Fn14 includes both apoptosis and necrosis and can be blocked by an anti-TWEAK antibody, CARL-1. Fn14 is expressed on HUVEC and in some cancer tissues but not on freshly isolated PBMCs. Fn14 mRNA expression has been identified during liver regeneration. Applications Reported: This ITEM-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ITEM-1 antibody has been pre-diluted and tested by flow cytometric analysis of human Fn14 transfected L5178Y cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
TWEAK is a member of the TNF family and was first described as a weak inducer of apoptosis in demonstrate that this relatively small molecule is a fully transformed cell lines. Recently, a receptor for TWEAK was isolated by expression cloning from a HUVEC cell cDNA library. TweakR is a small protein belonging to TNF receptor super family. TweakR is capable of intiating a proliferative signal by binding to TRAF1, 2, and 3. TweakR mRNA is upregulated during tissue regeneration in the aorta and liver. It may also play a role in vascular cell migration and angiogenesis.Spezifikation
CD266 (TWEAK Receptor) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9NP84 | |
TNFRSF12A | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TNFRSF12A |
ITEM-1 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
51330 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |