Learn More
CD271 (NGF Receptor) Monoclonal Antibody (ME20.4), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marke: Affymetrix eBioscience 62-9400-42
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The monoclonal antibody ME20.4 recognizes CD271, a 75 kDa, type I transmembrane protein that belongs to the low-affinity neurotrophin receptor and tumor necrosis factor receptor superfamily. CD271 is expressed by neurons, Schwann cells, melanocytes, mesen
CD271 (NGF Receptor) Monoclonal antibody specifically detects CD271 (NGF Receptor) in Canine, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat samples. It is validated for Flow Cytometry.Spezifikation
CD271 (NGF Receptor) | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P07174, P08138, Q9Z0W1 | |
Ngfr | |
Affinity chromatography | |
RUO | |
100349095, 100525607, 101119064, 18053, 24596, 4804, 491071 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
ME20.4 | |
Super Bright 436 | |
Ngfr | |
CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16 | |
Mouse | |
100 Tests | |
Primary | |
Canine, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat | |
Antibody | |
IgG1 κ |