Learn More
CD278 (ICOS) Monoclonal Antibody (7E.17G9), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
133.00 € - 305.00 €
Spezifikation
Antigen | CD278 (ICOS) |
---|---|
Klon | 7E.17G9 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15903234
|
Affymetrix eBioscience
17-9942-80 |
25 μg |
133.00 €
25 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15913234
|
Affymetrix eBioscience
17-9942-82 |
100 μg |
305.00 €
100 Mikrogramm |
Verfügbar ab: 07-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells. Applications Reported: This 7E.17G9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 7E.17G9 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser
CD278 (ICOS) Monoclonal antibody specifically detects CD278 (ICOS) in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD278 (ICOS) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
ICOS | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ICOS |
7E.17G9 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
Q9WVS0 | |
54167 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |