Learn More
CD28 Monoclonal Antibody (10F3), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen CD2804
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD28 | |
Monoclonal | |
PE | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Mouse | |
purified | |
ASR | |
940 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
10F3 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P10747 | |
Cd28 | |
Human CD28 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |