missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, BUV496, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 564810
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry.
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry.Spezifikation
CD3 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimum conditions and unconjugated antibody and free BD Horizon BUV496 were removed. | |
IgG1 κ |
Flow Cytometry | |
UCHT1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
For Research Use Only.