missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, APC, Clone: UCHT1, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB6-APC-25Tests
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CD3 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD3D | |
Protein A/G Purified | |
RUO | |
915 | |
IgG1 κ |
Flow Cytometry | |
UCHT1 | |
APC | |
NP_001035741 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |