missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD30 Ligand/TNFSF8 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | CD30 Ligand/TNFSF8 |
---|---|
Klon | 116615 |
Verdünnung | Western Blot 1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Beschreibung
CD30 Ligand/TNFSF8 Monoclonal specifically detects CD30 Ligand/TNFSF8 in Human samples. It is validated for Western Blot.Spezifikation
CD30 Ligand/TNFSF8 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD153, CD153 antigen, CD30 antigen ligand, CD30L, CD30-L, CD30LCD30 ligand, CD30LGMGC138144, TNFSF8, tumor necrosis factor (ligand) superfamily, member 8, tumor necrosis factor ligand superfamily member 8 | |
TNFSF8 | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
116615 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P32971 | |
944 | |
Mouse myeloma cell line NS0-derived recombinant human CD30 Ligand/TNFSF8 Gln63-Asp234 Accession # P32971 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD30 Ligand/TNFSF8 in direct ELISAs and Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse CD30 Ligand is observed. |
For Research Use Only