Learn More
CD4 Monoclonal Antibody (SK3 (SK-3)), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
102.00 € - 236.00 €
Spezifikation
Antigen | CD4 |
---|---|
Klon | SK3 (SK-3) |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Alexa Fluor 700 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16311024
|
Affymetrix eBioscience
56-0047-41 |
25 Tests |
102.00 €
25 Stück |
Verfügbar ab: 16-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16321024
|
Affymetrix eBioscience
56-0047-42 |
100 Tests |
236.00 €
100 Stück |
Verfügbar ab: 16-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: The SK3 monoclonal antibody reacts with human CD4, a 59-kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and at low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes. Applications Reported: This SK3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SK3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD4 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
Human | |
920 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
SK3 (SK-3) | |
Monoclonal | |
Liquid | |
RUO | |
P01730 | |
CD4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD4 |