missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse, FITC, Clone: SK3, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio QAB10-F-500Tests
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CD4 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD4 | |
Protein A/G Purified | |
RUO | |
920 | |
IgG1 κ |
Flow Cytometry | |
SK3 | |
FITC | |
NP_001181945 | |
Mouse | |
500 Tests | |
Primary | |
Store at 2-4°C. |