missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat anti-Mouse, BB700, Clone: RM4-5, BD Horizon™
Rat Monoclonal Antibody
Marke: BD Biosciences 566407
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD4 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
RM4-5 | |
Brilliant Blue 700 (BB700) | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |