Learn More
Invitrogen™ CD42a Monoclonal Antibody (GR-P), FITC, eBioscience™

Beschreibung
Description: This GR-P monoclonal antibody reacts with human CD42a, which is also known as glycoprotein IX (GPIX). CD42a is a 22-kDa transmembrane protein that is constitutively expressed on platelets and megakaryocytes. CD42a associates with CD42b, CD42c, and CD42d to form the GPIb-V-IX complex, which binds the von Willebrand factor, an endothelial adhesion protein, to mediate platelet adhesion and activation. This binding results in platelet aggregation at sites of vascular injury. This receptor complex has also been demonstrated to bind thrombin, factors XI and XII, P-selectin, and Mac-1. Finally, a point mutation within the leucine-rich motif of CD42a has been linked to Bernard-Soulier syndrome. Applications Reported: This GR-P antibody has been reported for use in flow cytometric analysis. Applications Tested: This GR-P antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Spezifikation
Spezifikation
| Antigen | CD42a |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | GR-P |
| Konzentration | 5 μL/Test |
| Konjugat | FITC |
| Zusammensetzung | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gen | GP9 |
| Gen-Zugriffsnummer | P14770 |
| Gen-Alias | CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.