missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD43 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | CD43/Sialophorin |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Anwendungen | Western Blot, Flow Cytometry, CyTOF |
Klassifikation | Polyclonal |
Beschreibung
CD43/Sialophorin Polyclonal specifically detects CD43/Sialophorin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Spezifikation
CD43/Sialophorin | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD43, CD43 antigen, CD43), Galactoglycoprotein, GALGP, GPL115, Leukocyte sialoglycoprotein, Leukosialin, LSN, Ly-48, Sialophorin, sialophorin (gpL115, leukosialin, CD43), SPN | |
SPN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
P16150 | |
6693 | |
Mouse myeloma cell line NS0-derived recombinant human CD43 Ser20-Arg253 Accession # P16150 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CD43 in direct ELISAs and Western blots. |
For Research Use Only