Learn More
ACROBiosystems CD47: SIRP alpha, Biotinylated, Inhibitor Screening ELISA Assay Pair
This pair is useful for screening for inhibitors of human CD47 binding to human SIRP alpha
737.89 €
Spezifikation
Produkttyp | Inhibitory Kit |
---|---|
Kitinhalt | This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new CD47 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human CD47 and in |
Versandbedingung | RT |
Lagerungsbedingungen | -20°C, aviod light |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16407499
|
ACROBiosystems
EP-102-96TESTS |
96 Tests |
737.89 €
96 Tests |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Immune checkpoint pathway is a focal point of today’s cancer research. CD47 is one of the best characterized checkpoint proteins. CD47 binding to SIRP alpha aids in tumor evasion of the immune system. Therefore, the pharmaceutical inhibition of CD47 has been considered a promising strategy by many oncologists.Spezifikation
Inhibitory Kit | |
This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new CD47 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human CD47 and in | |
RT | |
-20°C, aviod light |