Learn More
ACROBiosystems CD47: SIRP alpha, Biotinylated, Inhibitor Screening ELISA Assay Pair
This pair is useful for screening for inhibitors of human CD47 binding to human SIRP alpha
737.89 €
Spezifikation
Produkttyp Inhibitory Kit
Kitinhalt This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new CD47 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human CD47 and in
Versandbedingung RT
Lagerungsbedingungen -20°C, aviod light
Mehr Spezifikationen ansehen
Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16407499
Unterlagen ansehen Aktionsdetails
ACROBiosystems
EP-102-96TESTS
96 Tests
737.89 €
96 Tests
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 29-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Immune checkpoint pathway is a focal point of today’s cancer research. CD47 is one of the best characterized checkpoint proteins. CD47 binding to SIRP alpha aids in tumor evasion of the immune system. Therefore, the pharmaceutical inhibition of CD47 has been considered a promising strategy by many oncologists.
Spezifikation
Spezifikation
Inhibitory Kit
This inhibitor screening ELISA pair is designed to facilitate the identification and characterization of new CD47 pathway inhibitors. This assay employs a simple colorimetric ELISA platform, which measures the binding between immobilized human CD47 and in
RT
-20°C, aviod light