Learn More
CD56 (NCAM) Monoclonal Antibody (TULY56), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
324.00 €
Spezifikation
Antigen | CD56 (NCAM) |
---|---|
Klon | TULY56 |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16331084
|
Affymetrix eBioscience
61-0566-42 |
100 Tests |
324.00 €
100 Stück |
Verfügbar ab: 17-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Description: This TULY56 monoclonal antibody reacts with human CD56, also known as Neural Cell Adhesion Molecule (NCAM). CD56 is a highly glycosylated transmembrane molecule expressed by neurons and plays a role in the homotypic adhesion of neural cells. In the hematopoietic system, CD56 is expressed on NK cells and a subset of T cells referred to as NKT cells. Staining with TULY56 does not block binding of CMSSB, suggesting that the two antibodies recognize different epitopes. Additionally, TULY56 performs better after fixation and permeabilization than CMSSB. The TULY56 monoclonal antibody crossreacts with Rhesus macaque. Applications Reported: This TULY56 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TULY56 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PE-eFluor 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.
CD56 (NCAM) Monoclonal antibody specifically detects CD56 (NCAM) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Spezifikation
CD56 (NCAM) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P13591 | |
IgG1 κ | |
4° C, store in dark, DO NOT FREEZE! |
TULY56 | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human, Non-human Primate, Rhesus Monkey | |
4684, 693789 | |
Primary | |
Antibody |