missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD59b Antibody, R&D Systems™
Goat Polyclonal Antibody
N.V.
Spezifikation
Antigen | CD59b |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Beschreibung
CD59b Polyclonal specifically detects CD59b in Mouse samples. It is validated for Western Blot.Spezifikation
CD59b | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
966 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD59b Leu24-Asn104 Accession # P58019 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse CD59b in direct ELISAs and Western blots. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P58019 | |
Cd59b | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only