missing translation for 'onlineSavingsMsg'
Learn More

CD6 Mouse anti-Human, Clone: 123119, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication

N.V.

Spezifikation

Antigen CD6
Klon 123119
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD6 His18-Glu398 Accession # CAA43306
Gen-ID (Entrez) 923
Gen-Zugriffsnummer CAA43306
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

CD6 Monoclonal specifically detects CD6 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

CD6
Mouse myeloma cell line NS0-derived recombinant human CD6 His18-Glu398 Accession # CAA43306
CAA43306
Reconstitute at 0.5 mg/mL in sterile PBS.
Mouse
IgG1
Protein A or G purified from hybridoma culture supernatant
RUO
123119
923
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human CD6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD6 is observed.
Unconjugated
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only