Learn More
CD63 Antibody (LAMP3/9191R), mFluor Violet 610 SE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | CD63 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | LAMP3/9191R |
| Konjugat | mFluor Violet 610 SE |
| Zusammensetzung | 50 mM Sodium Borate |
| Gen-Alias | CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fragment (around aa100-200) of human CD63 protein (exact sequence is proprietary) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |