missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD63 Chimeric Recombinant Rabbit Monoclonal Antibody (NK-1-C3)
GREENER_CHOICE

Artikelnummer. 17921461
Change view
Click to view available options
Menge:
200 μg
Packungsgröße:
200 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17921461 200 μg 200 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17921461 Lieferant Invitrogen™ Lieferanten-Nr. MA548275

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody recognizes CD63. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation and plays a role in the activation of cellular signaling cascades.

CD63, also known as LAMP-3, is a glycoprotein belonging to the tetraspanin family, primarily located in late endosomes, lysosomes, and secretory vesicles across various cell types. It is expressed intracellularly in resting platelets and basophils, but upon activation, CD63 translocates to the plasma membrane, making it a widely used marker for basophil activation. Unlike basophils, mast cells can expose CD63 without activation. CD63 is expressed on activated platelets, monocytes, macrophages, and granulocytes, as well as endothelial cells, fibroblasts, and smooth muscle cells. It interacts with integrins (such as VLA-3 and VLA-6) and TIMP-1, playing roles in mediating allergic responses, phagocytosis, and cell migration. Additionally, CD63 is involved in the regulation of H/K-ATPase trafficking and ROMK1 channels, and serves as a T-cell costimulation molecule. CD63 expression can be used to predict prognosis in early-stage carcinomas and is identical to the melanoma-associated antigen ME491 and the platelet antigen PTLGP40. Diseases associated with CD63 dysfunction include melanoma and Hermansky-Pudlak Syndrome, highlighting its significance in both immune responses and disease processes.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CD63 Chimeric
Anwendungen Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Klassifikation Recombinant Monoclonal
Klon NK-1-C3
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.02% ProClin 300
Gen CD63
Gen-Zugriffsnummer P08962, P41731
Gen-Alias C75951; CD 63; Cd63; CD63 antigen; CD63 antigen (melanoma 1 antigen); Cd63 molecule; CD63 protein; granulophysin; LAMP-3; Lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; mast cell antigen AD1; ME491; melanoma 1 antigen; melanoma-associated antigen ME491; melanoma-associated antigen MLA1; MLA1; Ocular melanoma-associated antigen; OMA81H; Tetraspanin-30; TSPAN30; tspan-30
Gensymbole CD63
Wirtsspezies Rabbit
Immunogen This antibody was raised by immunizing BALB/c mice with a plasma membrane preparation of the human melanoma MeWo cells.
Reinigungsverfahren Protein A
Menge 200 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 12512, 967
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.