missing translation for 'onlineSavingsMsg'
Learn More

CD69 Monoclonal Antibody (FN50), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

134.00 € - 343.00 €

Spezifikation

Antigen CD69
Klon FN50
Konzentration 5 μL/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15903394
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
62-0699-42
100 Tests
343.00 €
100 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
15993384
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
62-0699-41
25 Tests Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

Description: The FN50 monoclonal antibody reacts with human CD69, also known as very early activation antigen (VEA). CD69 is approximately 30 kDa and is expressed on the cell-surface as a disulfide-linked dimer. CD69 is rapidly upregulated upon activation and expressed on lymphocytes, monocytes and platelets. Applications Reported: This FN50 antibody has been reported for use in flow cytometric analysis. Applications Tested: This FN50 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.

CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD69
5 μL/Test
Monoclonal
Liquid
RUO
Q07108
CD69
Primary
4° C, store in dark, DO NOT FREEZE!
CD69
FN50
Flow Cytometry
Super Bright 436
Mouse
Human
969
IgG1 κ
Affinity chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen