missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD73 Monoclonal Antibody (eBioTY/11.8 (TY/11.8)), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 63-0731-80
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
CD73 Monoclonal antibody specifically detects CD73 in Mouse samples. It is validated for Flow CytometrySpezifikation
CD73 | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q61503 | |
NT5E | |
Affinity chromatography | |
RUO | |
23959 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
eBioTY/11.8 (TY/11.8) | |
Super Bright 600 | |
NT5E | |
2210401F01Rik; 5'-NT; 5'-nucleotidase; AI447961; CALJA; CD73; E5NT; Ecto-5'-nucleotidase; eN; eNT; NT; Nt5; Nt5e; NTE | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |