missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81 / TAPA-1 Mouse, Clone: 1.3.3.22, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 975-MSM1-P1
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
CD81 / TAPA-1 | |
Cell surface | |
1.3.3.22 | |
Unconjugated | |
NP_001284578 | |
Mouse | |
26kDa | |
Protein A/G Purified | |
RUO | |
975 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD81 | |
B-Cell line derived from a Burkitt lymphoma | |
Ramos, Daudi, Raji, U266, HUT-78, HUT-102, Jurkat, HL-60, KG1a, A293, A431, MG63, and human lymphocytes. Tonsil or Liver. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |