missing translation for 'onlineSavingsMsg'
Learn More

CD84/SLAMF5 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

N.V.

Spezifikation

Antigen CD84/SLAMF5
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD84/SLAMF5 Lys22-Arg220 Accession # Q9UIB8.1
Gen-ID (Entrez) 8832
Gen-Zugriffsnummer Q9UIB8.1
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

CD84/SLAMF5 Polyclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spezifikation

Spezifikation

CD84/SLAMF5
8832
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human CD84/SLAMF5 in direct ELISAs and Western blots.
Unconjugated
Western Blot, Flow Cytometry, CyTOF
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Mouse myeloma cell line NS0-derived recombinant human CD84/SLAMF5 Lys22-Arg220 Accession # Q9UIB8.1
Q9UIB8.1
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only