Learn More
CD86 Mouse anti-Human, FITC, Clone: BU63, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech FITC-65165-100TESTS
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD86 (also known as B7.2) is a costimulatory molecule belonging to the immunoglobulin superfamily. Primarily expressed on antigen-presenting cells (APCs), including B cells, dendritic cells, and macrophages, CD86 is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of CD86 with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of CD86 with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response.
Spezifikation
CD86 | |
Monoclonal | |
20 μL/Test | |
Mouse | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
BU63 | |
FITC | |
100 Tests | |
942 | |
Liquid |